Product References
MiR-451a and let-7i-5p loaded extracellular vesicles attenuate heme-induced inflammation in hiPSC-derived endothelial cells.
Frontiers in immunology
Thomas JJ,Harp KO,Bashi A,Hood JL,Botchway F,Wilson MD,Thompson WE,Stiles JK,Driss A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Wed Jan 11 00:00:00 EST 2023
Angiogenic stem cell delivery platform to augment post-infarction neovasculature and reverse ventricular remodeling.
Scientific reports
Shin HS,Thakore A,Tada Y,Pedroza AJ,Ikeda G,Chen IY,Chan D,Jaatinen KJ,Yajima S,Pfrender EM,Kawamura M,Yang PC,Wu JC,Appel EA,Fischbein MP,Woo Y,Shudo Y
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Thu Oct 20 00:00:00 EDT 2022
Autophagy modulates endothelial junctions to restrain neutrophil diapedesis during inflammation.
Immunity
Reglero-Real N,Pérez-Gutiérrez L,Yoshimura A,Rolas L,Garrido-Mesa J,Barkaway A,Pickworth C,Saleeb RS,Gonzalez-Nuñez M,Austin-Williams SN,Cooper D,Vázquez-Martínez L,Fu T,De Rossi G,Golding M,Voisin MB,Boulanger CM,Kubota Y,Muller WA,Tooze SA,Nightingale TD,Collinson L,Perretti M,Aksoy E,Nourshargh S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunocytochemistry
Tue Sep 14 00:00:00 EDT 2021
Simultaneously targeting cancer-associated fibroblasts and angiogenic vessel as a treatment for TNBC.
The Journal of experimental medicine
Sharma M,Turaga RC,Yuan Y,Satyanarayana G,Mishra F,Bian Z,Liu W,Sun L,Yang J,Liu ZR
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunocytochemistry
Mon Apr 05 00:00:00 EDT 2021
Simultaneously targeting cancer-associated fibroblasts and angiogenic vessel as a treatment for TNBC.
The Journal of experimental medicine
Sharma M,Turaga RC,Yuan Y,Satyanarayana G,Mishra F,Bian Z,Liu W,Sun L,Yang J,Liu ZR
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunocytochemistry
Mon Apr 05 00:00:00 EDT 2021
Modeling of endothelial cell dysfunction using human induced pluripotent stem cells derived from patients with end-stage renal disease.
Kidney research and clinical practice
Kim KW,Shin YJ,Kim BM,Cui S,Ko EJ,Lim SW,Yang CW,Chung BH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunocytochemistry
Wed Dec 01 00:00:00 EST 2021
Expression and role of interleukin-1β and associated biomarkers in deep vein thrombosis.
Experimental and therapeutic medicine
Pai RZ,Fang Q,Tian G,Zhu B,Ge X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Wed Dec 01 00:00:00 EST 2021
Injectable hydrogel with MSNs/microRNA-21-5p delivery enables both immunomodification and enhanced angiogenesis for myocardial infarction therapy in pigs.
Science advances
Li Y,Chen X,Jin R,Chen L,Dang M,Cao H,Dong Y,Cai B,Bai G,Gooding JJ,Liu S,Zou D,Zhang Z,Yang C
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunohistochemistry
Mon Feb 01 00:00:00 EST 2021
Injectable hydrogel with MSNs/microRNA-21-5p delivery enables both immunomodification and enhanced angiogenesis for myocardial infarction therapy in pigs.
Science advances
Li Y,Chen X,Jin R,Chen L,Dang M,Cao H,Dong Y,Cai B,Bai G,Gooding JJ,Liu S,Zou D,Zhang Z,Yang C
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunohistochemistry
Mon Feb 01 00:00:00 EST 2021
A Human Skeletal Muscle Atlas Identifies the Trajectories of Stem and Progenitor Cells across Development and from Human Pluripotent Stem Cells.
Cell stem cell
Xi H,Langerman J,Sabri S,Chien P,Young CS,Younesi S,Hicks M,Gonzalez K,Fujiwara W,Marzi J,Liebscher S,Spencer M,Van Handel B,Evseenko D,Schenke-Layland K,Plath K,Pyle AD
11-0319-42 was used in Flow Cytometry to profile human embryonic, fetal and postnatal skeletal muscle tissues to develop a roadmap of muscle ontogeny.
Thu Jul 02 00:00:00 EDT 2020
Placental endovascular extravillous trophoblasts (enEVTs) educate maternal T-cell differentiation along the maternal-placental circulation.
Cell proliferation
Ma Y,Yang Q,Fan M,Zhang L,Gu Y,Jia W,Li Z,Wang F,Li YX,Wang J,Li R,Shao X,Wang YL
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Fri May 01 00:00:00 EDT 2020
CD26 Identifies a Subpopulation of Fibroblasts that Produce the Majority of Collagen during Wound Healing in Human Skin.
The Journal of investigative dermatology
Worthen CA,Cui Y,Orringer JS,Johnson TM,Voorhees JJ,Fisher GJ
11-0319 was used in Flow cytometry/Cell sorting to provide a foundation for targeting CD26+ fibroblasts to modulate wound healing in human skin.
Tue Dec 01 00:00:00 EST 2020
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
Aging affects responsiveness of peripheral blood mononuclear cells to immunosuppression of periodontal ligament stem cells.
The Journal of international medical research
Li X,Zhang B,Wang H,Zhao X,Zhang Z,Ding G,Wei F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2020
Aging affects responsiveness of peripheral blood mononuclear cells to immunosuppression of periodontal ligament stem cells.
The Journal of international medical research
Li X,Zhang B,Wang H,Zhao X,Zhang Z,Ding G,Wei F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
11-0319-42 was used in Flow cytometry/Cell sorting to investigate the role of Pluronic F-127 (PF-127) hydrogel plus antioxidant sodium ascorbyl phosphate (SAP) in enhancing Wharton's jelly mesenchymal stem cell (WJMSC)-mediated effectiveness on full-thickness skin wound healing in mice.
Fri Apr 03 00:00:00 EDT 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
11-0319-42 was used in Flow cytometry/Cell sorting to investigate the role of Pluronic F-127 (PF-127) hydrogel plus antioxidant sodium ascorbyl phosphate (SAP) in enhancing Wharton's jelly mesenchymal stem cell (WJMSC)-mediated effectiveness on full-thickness skin wound healing in mice.
Fri Apr 03 00:00:00 EDT 2020
Developing Human Skin Contains Lymphocytes Demonstrating a Memory Signature.
Cell reports. Medicine
Dhariwala MO,Karthikeyan D,Vasquez KS,Farhat S,Weckel A,Taravati K,Leitner EG,Clancy S,Pauli M,Piper ML,Cohen JN,Ashouri JF,Lowe MM,Rosenblum MD,Scharschmidt TC
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Tue Nov 17 00:00:00 EST 2020
Both Human Hematoma Punctured from Pelvic Fractures and Serum Increase Muscle Resident Stem Cells Response to BMP9: A Multivariate Statistical Approach.
Journal of clinical medicine
Alinejad Y,Lauzon MA,Grenier G,Balg F,Faucheux N
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Sun Apr 19 00:00:00 EDT 2020
Functionally specialized human CD4+ T-cell subsets express physicochemically distinct TCRs.
eLife
Kasatskaya SA,Ladell K,Egorov ES,Miners KL,Davydov AN,Metsger M,Staroverov DB,Matveyshina EK,Shagina IA,Mamedov IZ,Izraelson M,Shelyakin PV,Britanova OV,Price DA,Chudakov DM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Tue Dec 08 00:00:00 EST 2020
Vascular Tumor Recapitulated in Endothelial Cells from hiPSCs Engineered to Express the SERPINE1-FOSB Translocation.
Cell reports. Medicine
van IJzendoorn DGP,Salvatori DCF,Cao X,van den Hil F,Briaire-de Bruijn IH,de Jong D,Mei H,Mummery CL,Szuhai K,Bovée JVMG,Orlova VV
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Tue Dec 22 00:00:00 EST 2020
Hyaluronan-CD44 interactions mediate contractility and migration in periodontal ligament cells.
Cell adhesion & migration
Al-Rekabi Z,Fura AM,Juhlin I,Yassin A,Popowics TE,Sniadecki NJ
11-0319 was used in Flow cytometry/Cell sorting to determine the effect of exogenous hyaluronan on periodontal ligament cells.
Sun Dec 01 00:00:00 EST 2019
Mesenchymal stromal cells lower platelet activation and assist in platelet formation in vitro.
JCI insight
Mendelson A,Strat AN,Bao W,Rosston P,Fallon G,Ohrn S,Zhong H,Lobo C,An X,Yazdanbakhsh K
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunohistochemistry
Thu Aug 22 00:00:00 EDT 2019
Matrine inhibits the development and progression of ovarian cancer by repressing cancer associated phosphorylation signaling pathways.
Cell death & disease
Zhang X,Hou G,Liu A,Xu H,Guan Y,Wu Y,Deng J,Cao X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunohistochemistry
Thu Oct 10 00:00:00 EDT 2019
Spatial and Single-Cell Transcriptional Profiling Identifies Functionally Distinct Human Dermal Fibroblast Subpopulations.
The Journal of investigative dermatology
Philippeos C,Telerman SB,Oulès B,Pisco AO,Shaw TJ,Elgueta R,Lombardi G,Driskell RR,Soldin M,Lynch MD,Watt FM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
CRISPR/Cas9‑mediated hypoxia inducible factor‑1α knockout enhances the antitumor effect of transarterial embolization in hepatocellular carcinoma.
Oncology reports
Liu Q,Fan D,Adah D,Wu Z,Liu R,Yan QT,Zhang Y,Du ZY,Wang D,Li Y,Bao SY,Liu LP
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunohistochemistry (Paraffin)
Thu Nov 01 00:00:00 EDT 2018
Expansion of functional personalized cells with specific transgene combinations.
Nature communications
Lipps C,Klein F,Wahlicht T,Seiffert V,Butueva M,Zauers J,Truschel T,Luckner M,Köster M,MacLeod R,Pezoldt J,Hühn J,Yuan Q,Müller PP,Kempf H,Zweigerdt R,Dittrich-Breiholz O,Pufe T,Beckmann R,Drescher W,Riancho J,Sañudo C,Korff T,Opalka B,Rebmann V,Göthert JR,Alves PM,Ott M,Schucht R,Hauser H,Wirth D,May T
11-0319 was used in Flow cytometry/Cell sorting to develop a novel strategy for rapid, functional, personalised cell expansion.
Thu Mar 08 00:00:00 EST 2018
Expansion of functional personalized cells with specific transgene combinations.
Nature communications
Lipps C,Klein F,Wahlicht T,Seiffert V,Butueva M,Zauers J,Truschel T,Luckner M,Köster M,MacLeod R,Pezoldt J,Hühn J,Yuan Q,Müller PP,Kempf H,Zweigerdt R,Dittrich-Breiholz O,Pufe T,Beckmann R,Drescher W,Riancho J,Sañudo C,Korff T,Opalka B,Rebmann V,Göthert JR,Alves PM,Ott M,Schucht R,Hauser H,Wirth D,May T
11-0319 was used in Flow cytometry/Cell sorting to develop a novel strategy for rapid, functional, personalised cell expansion.
Thu Mar 08 00:00:00 EST 2018
Prostaglandin E2 Is Required for BMP4-Induced Mesoderm Differentiation of Human Embryonic Stem Cells.
Stem cell reports
Zhang B,He L,Liu Y,Zhang J,Zeng Q,Wang S,Fan Z,Fang F,Chen L,Lv Y,Xi J,Yue W,Li Y,Pei X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Tue Mar 13 00:00:00 EDT 2018
Exosomes secreted by adipose-derived mesenchymal stem cells regulate type I collagen metabolism in fibroblasts from women with stress urinary incontinence.
Stem cell research & therapy
Liu X,Wang S,Wu S,Hao Q,Li Y,Guo Z,Wang W
11-0319 was used in Flow cytometry/Cell sorting to evaluate the effects of exomes secreted by adipose-derived mesenchymal stem cells on collagen metabolism in cultured fibroblasts from women with stress urinary incontinence.
Wed Jun 13 00:00:00 EDT 2018
MMP-2 and MMP-14 Silencing Inhibits VEGFR2 Cleavage and Induces the Differentiation of Porcine Adipose-Derived Mesenchymal Stem Cells to Endothelial Cells.
Stem cells translational medicine
Almalki SG,Llamas Valle Y,Agrawal DK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Mon May 01 00:00:00 EDT 2017
ERK signaling is required for VEGF-A/VEGFR2-induced differentiation of porcine adipose-derived mesenchymal stem cells into endothelial cells.
Stem cell research & therapy
Almalki SG,Agrawal DK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Fri May 12 00:00:00 EDT 2017
Endothelial and smooth muscle cells derived from human cardiac explants demonstrate angiogenic potential and suitable for design of cell-containing vascular grafts.
Journal of translational medicine
Zakharova IS,Zhiven' MK,Saaya SB,Shevchenko AI,Smirnova AM,Strunov A,Karpenko AA,Pokushalov EA,Ivanova LN,Makarevich PI,Parfyonova YV,Aboian E,Zakian SM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunofluorescence
Fri Mar 03 00:00:00 EST 2017
TGF-β reduces DNA ds-break repair mechanisms to heighten genetic diversity and adaptability of CD44+/CD24- cancer cells.
eLife
Pal D,Pertot A,Shirole NH,Yao Z,Anaparthy N,Garvin T,Cox H,Chang K,Rollins F,Kendall J,Edwards L,Singh VA,Stone GC,Schatz MC,Hicks J,Hannon GJ,Sordella R
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Mon Jan 16 00:00:00 EST 2017
Increased infiltration and tolerised antigen-specific CD8+ TEM cells in tumor but not peripheral blood have no impact on survival of HCMV+ glioblastoma patients.
Oncoimmunology
Bahador M,Gras Navarro A,Rahman MA,Dominguez-Valentin M,Sarowar S,Ulvestad E,Njølstad G,Lie SA,Kristoffersen EK,Bratland E,Chekenya M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Targeted Disruption of TCF12 Reveals HEB as Essential in Human Mesodermal Specification and Hematopoiesis.
Stem cell reports
Li Y,Brauer PM,Singh J,Xhiku S,Yoganathan K,Zúñiga-Pflücker JC,Anderson MK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Tue Sep 12 00:00:00 EDT 2017
CXCL12/CXCR4 activation by cancer-associated fibroblasts promotes integrin β1 clustering and invasiveness in gastric cancer.
International journal of cancer
Izumi D,Ishimoto T,Miyake K,Sugihara H,Eto K,Sawayama H,Yasuda T,Kiyozumi Y,Kaida T,Kurashige J,Imamura Y,Hiyoshi Y,Iwatsuki M,Iwagami S,Baba Y,Sakamoto Y,Miyamoto Y,Yoshida N,Watanabe M,Takamori H,Araki N,Tan P,Baba H
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2016
Oxidized macrophage migration inhibitory factor is a potential new tissue marker and drug target in cancer.
Oncotarget
Schinagl A,Thiele M,Douillard P,Völkel D,Kenner L,Kazemi Z,Freissmuth M,Scheiflinger F,Kerschbaumer RJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunofluorescence
Tue Nov 08 00:00:00 EST 2016
Acidic pH reduces VEGF-mediated endothelial cell responses by downregulation of VEGFR-2; relevance for anti-angiogenic therapies.
Oncotarget
Faes S,Uldry E,Planche A,Santoro T,Pythoud C,Demartines N,Dormond O
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunofluorescence
Tue Dec 27 00:00:00 EST 2016
Efficient gene disruption in cultured primary human endothelial cells by CRISPR/Cas9.
Circulation research
Abrahimi P,Chang WG,Kluger MS,Qyang Y,Tellides G,Saltzman WM,Pober JS
11-0319 was used in Flow cytometry/Cell sorting to demonstrate that the CRISPR/Cas9 modification of epithelial cells is a powerful platform for vascular research and for regenerative medicine.
Fri Jul 03 00:00:00 EDT 2015
Mesenchymal stromal cells form vascular tubes when placed in fibrin sealant and accelerate wound healing in vivo.
Biomaterials
Mendez JJ,Ghaedi M,Sivarapatna A,Dimitrievska S,Shao Z,Osuji CO,Steinbacher DM,Leffell DJ,Niklason LE
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunofluorescence
Sun Feb 01 00:00:00 EST 2015
Mesenchymal stromal cells form vascular tubes when placed in fibrin sealant and accelerate wound healing in vivo.
Biomaterials
Mendez JJ,Ghaedi M,Sivarapatna A,Dimitrievska S,Shao Z,Osuji CO,Steinbacher DM,Leffell DJ,Niklason LE
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Immunofluorescence
Sun Feb 01 00:00:00 EST 2015
Activation of the Wnt/planar cell polarity pathway is required for pericyte recruitment during pulmonary angiogenesis.
The American journal of pathology
Yuan K,Orcholski ME,Panaroni C,Shuffle EM,Huang NF,Jiang X,Tian W,Vladar EK,Wang L,Nicolls MR,Wu JY,de Jesus Perez VA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Thu Jan 01 00:00:00 EST 2015
Human esophageal myofibroblasts secrete proinflammatory cytokines in response to acid and Toll-like receptor 4 ligands.
American journal of physiology. Gastrointestinal and liver physiology
Gargus M,Niu C,Vallone JG,Binkley J,Rubin DC,Shaker A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2015
Relationship of tumor marker CA125 and ovarian tumor stem cells: preliminary identification.
Journal of ovarian research
Zhang H,Yang Y,Wang Y,Gao X,Wang W,Liu H,He H,Liang Y,Pan K,Wu H,Shi J,Xue H,Liang L,Cai Z,Fan Y,Zhang Y
11-0319 was used in Flow cytometry/Cell sorting to study the prospective association between CA125 and tumorigenic ovarian cancer cells, showing that CA125+ cells could form new tumours and may be ovarian cancer stem cells.
Sat Mar 28 00:00:00 EDT 2015
Reprogramming human endothelial cells to haematopoietic cells requires vascular induction.
Nature
Sandler VM,Lis R,Liu Y,Kedem A,James D,Elemento O,Butler JM,Scandura JM,Rafii S
11-0319 was used in Flow cytometry/Cell sorting to investigate the role of inductive cues from the vascular niche in coordinating and sustaining haematopoietic specification.
Thu Jul 17 00:00:00 EDT 2014
Vascular progenitors from cord blood-derived induced pluripotent stem cells possess augmented capacity for regenerating ischemic retinal vasculature.
Circulation
Park TS,Bhutto I,Zimmerlin L,Huo JS,Nagaria P,Miller D,Rufaihah AJ,Talbot C,Aguilar J,Grebe R,Merges C,Reijo-Pera R,Feldman RA,Rassool F,Cooke J,Lutty G,Zambidis ET
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Tue Jan 21 00:00:00 EST 2014
The CD14+CD16+ inflammatory monocyte subset displays increased mitochondrial activity and effector function during acute Plasmodium vivax malaria.
PLoS pathogens
Antonelli LR,Leoratti FM,Costa PA,Rocha BC,Diniz SQ,Tada MS,Pereira DB,Teixeira-Carvalho A,Golenbock DT,Gonçalves R,Gazzinelli RT
11-0319 was used in Flow cytometry/Cell sorting to study inflammatory monocyte subsets displaying increased mitochondrial activity and effector function during malaria.
Mon Sep 01 00:00:00 EDT 2014
Association of obesity and circulating adipose stromal cells among breast cancer survivors.
Molecular biology reports
Ghosh S,Hughes D,Parma DL,Ramirez A,Li R
11-0319 was used in Flow cytometry/Cell sorting to suggest that circulating ASCs can serve as a potential biomarker for future studies of the impacts of obesity and physical activity on breast cancer recurrence and survival.
Thu May 01 00:00:00 EDT 2014
Hypoxia-cultured human adipose-derived mesenchymal stem cells are non-oncogenic and have enhanced viability, motility, and tropism to brain cancer.
Cell death & disease
Feng Y,Zhu M,Dangelmajer S,Lee YM,Wijesekera O,Castellanos CX,Denduluri A,Chaichana KL,Li Q,Zhang H,Levchenko A,Guerrero-Cazares H,Quiñones-Hinojosa A
Thu Dec 11 00:00:00 EST 2014
Efficient and simultaneous generation of hematopoietic and vascular progenitors from human induced pluripotent stem cells.
Cytometry. Part A : the journal of the International Society for Analytical Cytology
Park TS,Zimmerlin L,Zambidis ET
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Tue Jan 01 00:00:00 EST 2013
Detection and validation of circulating endothelial cells, a blood-based diagnostic marker of acute myocardial infarction.
PloS one
Li C,Wu Q,Liu B,Yao Y,Chen Y,Zhang H,Wang C,Cao J,Ge S
11-0319 was used in Flow cytometry/Cell sorting to investigate the diagnostic and prognostic value of circulating endothelial cell monitoring in acute myocardial infarction patients.
Tue Sep 03 00:00:00 EDT 2013
Efficient direct reprogramming of mature amniotic cells into endothelial cells by ETS factors and TGFβ suppression.
Cell
Ginsberg M,James D,Ding BS,Nolan D,Geng F,Butler JM,Schachterle W,Pulijaal VR,Mathew S,Chasen ST,Xiang J,Rosenwaks Z,Shido K,Elemento O,Rabbany SY,Rafii S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Fri Oct 26 00:00:00 EDT 2012
Efficient direct reprogramming of mature amniotic cells into endothelial cells by ETS factors and TGFβ suppression.
Cell
Ginsberg M,James D,Ding BS,Nolan D,Geng F,Butler JM,Schachterle W,Pulijaal VR,Mathew S,Chasen ST,Xiang J,Rosenwaks Z,Shido K,Elemento O,Rabbany SY,Rafii S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Fri Oct 26 00:00:00 EDT 2012
Potential factors influencing the development of thrombocytopenia and consumptive coagulopathy after genetically modified pig liver xenotransplantation.
Transplant international : official journal of the European Society for Organ Transplantation
Ekser B,Lin CC,Long C,Echeverri GJ,Hara H,Ezzelarab M,Bogdanov VY,Stolz DB,Enjyoji K,Robson SC,Ayares D,Dorling A,Cooper DK,Gridelli B
11-0319 was used in Flow cytometry/Cell sorting to reveal tissue factor exposure on recipient platelets and peripheral blood mononuclear cells as a potential key factor in the development of thrombocytopenia.
Wed Aug 01 00:00:00 EDT 2012
Cells expressing the C/EBPbeta isoform, LIP, engulf their neighbors.
PloS one
Abreu M,Sealy L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 11-0319-42) in Flow Cytometry
Thu Apr 11 00:00:00 EDT 2013